RAMCO AA-003-01 RTY Acid Neutralizer, 8 oz Bottle
Representative image only.
Mfr # AA-003-01 – Item # EW-08779-64
Protect employees, equipment, and the environment
PRODUCT OPTIONS
$49.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • DescriptionRTY Acid Neutralizer, 8 oz Bottle
More About this Item
RTY Neutralizer™ is a caustic solution used concurrently with ON GUARD™ Acid Detecting Paint or Acid Spot'r™. RTY Neutralizer™ has a pH 11.5 and can be used to neutralize an acid spill or leak. Use RTY Neutralizer™ to expedite the color transition of ON GUARD™ after an acid exposure. After there has been an acid leak on a surface that has been painted with ON GUARD™, isolate and repair the leak, and then irrigate the site with RTY Neutralizer™. Once the indicating red color changes back to golden yellow, rinse the site with water and ON GUARD™ is ready to detect the next leak. Always contain spills and leaks and dispose of all waste materials in accordance with all regulations.