Opto 22 E2 Digital Brain Pamux, 16-channel
Representative image only.
Opto 22 Mfr # B5 – Item # EW-68494-25
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigital Brain Pamux, 16-channel
More About this Item
These Pamux brain boards operate in high-speed, high-density Pamux distributed I/O systems. Pamux systems are ideal for high speed, low noise applications such as robotics and numerical control. The Pamux B4 is an addressable digital brain board that can control up to 32 I/O points. Up to 16 B4 boards can be linked together to operate up to 512 I/O points. The B4 features a 70-pin header connector that attach to a digital I/O mounting rack. The B5 is an addressable digital brain board that can control up to 16 I/O points and up to 32 boards can be linked on a single Pamux bus to control up to 512 I/O points. The B5 features a 50-pin female connector to attach to a mounting rack and two 50-pin male connectors to attach to the Pamux bus or a terminator board. The B6 is an addressable analog brain board that can control up to 16 I/O points and up to 32 B6 boards may be linked on a single Pamux bus to control up to 512 points. The B6 also includes a microprocessor that continually scans all I/O points and communicates updates to a dual-port RAM. The B6 features a 50-pin female connector to attach to a mounting rack and two 50-pin male connectors to attach to the Pamux bus or a terminator board.