Opto 22 MP240D4 DC Control SSR, 240 VAC, 4 A
Representative image only.
Opto 22 Mfr # MP240D4 – Item # EW-68474-31
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Load Rating240 VAC
  • Max Current (amps)4
  • DescriptionDC Control SSR, 240 VAC, 4 A
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
The MP Series packaging is designed with a minimum footprint to allow maximum relay density on the circuit board. All MP Series SSRs provide 4000 VRMS of optical isolation and provide a wide operating temperature range of -40°C to +100°C. MP Series SSRs are TTL compatible, with a 3-32 VDC control voltage. All MP Series SSRs feature a built-in snubber and a maximum zero voltage turn-on time of 1/2 cycle. All models are UL recognized and CSA certified.
Opto 22’s Printed Circuit Series allows OEMs to easily deploy solid-state relays on printed circuit boards with switch loads up to 4 amps.