Opto 22 SNAP-IAC-16 Digital Input Module, 16-ch, 90/140 VAC/VDC
Representative image only.
Opto 22 Mfr # SNAP-IAC-16 – Item # EW-20011-99
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Input TypeDigital
  • Number of Channels16
  • Max Voltage Output (VDC)140
  • Min Votlage Output (VDC)90
  • Max Voltage Output (VAC)140
  • Min Votlage Output (VAC)90
  • DescriptionDigital Input Module, 16-ch, 90/140 VAC/VDC
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
G4 I/O includes both digital and analog modules. Each G4 module offers high-density single-channel packaging. Each digital module includes an LED status indicator and is also available with manual switching for field wiring and control logic testing. Digital modules support 5, 15, or 24-volt logic. G4 digital output modules feature a top-mounted replaceable fuse. G4 analog modules are factory-calibrated, offer 12-bit resolution, and requires a single 24-volt power supply. All current loop modules include an isolated loop power supply for added flexibility and reduced instrumentation costs. Input types include voltage, current, thermocouple, RTD, ICTD, frequency, and velocity. Outputs include voltage, current, and time-proportional output. Factory Mutual (FM) approved versions of many G4 I/O modules are available. Please inquire for details on specific models.