Opto 22 SNAP-IDC-16 Digital Input Module, 16-ch, 10/32 VAC/VDC
Representative image only.
Opto 22 Mfr # SNAP-IDC-16 – Item # EW-20012-04
The SNAP PAC System is an integrated system of hardware and software for industrial control remote monitoring and data acquisition applications
  • Standalone controller suitable for larger and more complex applications
  • Up to 32 ioControl flowcharts running simultaneously
  • Two 10/100 Mbps Ethernet interfaces for host and I/O communication over standard Ethernet networks
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Input TypeDigital
  • Number of Channels16
  • Max Voltage Output (VDC)32
  • Min Votlage Output (VDC)10
  • DescriptionDigital Input Module, 16-ch, 10/32 VAC/VDC
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Standalone controller suitable for larger and more complex applications
  • Up to 32 ioControl flowcharts running simultaneously
  • Two 10/100 Mbps Ethernet interfaces for host and I/O communication over standard Ethernet networks
  • Limited Lifetime Warranty
More About this Item
S Series
One RS-485 serial port for connection to Opto 22 Mistic I/O units
Two RS-232 serial ports (one with full handshaking) for PPP host connection or direct connections to
serial devices
Multiple protocol support: TCP/IP, SNMP, SMTP, FTP, OptoMMP™
R Series
Convenient PAC package for small or distributed applications
Compact Rack-mounted industrial controller with I/O processor included
(PAC R-series)
Two independent 10/100 Mbps Ethernet network interfaces
One RS-232 port with handshaking suitable for modem communications using PPP
Full-featured control and HMI software included with purchase