Opto 22 SNAP-IDC5-SW Contact Status Input Module, 4-channel, normally open
Representative image only.
Opto 22 Mfr # SNAP-IDC5-SW – Item # EW-68475-30
The SNAP PAC System is an integrated system of hardware and software for industrial control remote monitoring and data acquisition applications
  • Standalone controller suitable for larger and more complex applications
  • Up to 32 ioControl flowcharts running simultaneously
  • Two 10/100 Mbps Ethernet interfaces for host and I/O communication over standard Ethernet networks
  • 2-1/2 Year Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Input TypeDigital
  • Number of Channels4
  • Output 1Switch: normally open
  • DescriptionContact Status Input Module, 4-channel, normally open
  • Warranty2-1/2 Years Warranty
Key Features
  • Standalone controller suitable for larger and more complex applications
  • Up to 32 ioControl flowcharts running simultaneously
  • Two 10/100 Mbps Ethernet interfaces for host and I/O communication over standard Ethernet networks
  • 2-1/2 Year Warranty
More About this Item
S Series
One RS-485 serial port for connection to Opto 22 Mistic I/O units
Two RS-232 serial ports (one with full handshaking) for PPP host connection or direct connections to
serial devices
Multiple protocol support: TCP/IP, SNMP, SMTP, FTP, OptoMMP™
R Series
Convenient PAC package for small or distributed applications
Compact Rack-mounted industrial controller with I/O processor included
(PAC R-series)
Two independent 10/100 Mbps Ethernet network interfaces
One RS-232 port with handshaking suitable for modem communications using PPP
Full-featured control and HMI software included with purchase