Opto 22 SNAP-LCM4 Controller for FactoryFloor Systems
Representative image only.
Opto 22 Mfr # SNAP-LCM4 – Item # EW-68476-13
  • 2-1/2 Year Warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionController for FactoryFloor Systems
  • Warranty2-1/2 Years Warranty
Key Features
  • 2-1/2 Year Warranty
More About this Item
SNAP controllers work with SNAP I/O systems to provide powerful distributed control in a small footprint. The SNAP-LCM4 is Opto 22’s fastest controller to date, ideal for control and communication to I/O systems, serial devices, motion controllers, and networks.
The SNAP-LCM4 features powerful communications capabilities including Ethernet, ARCNET, and serial. Serial ports provide an interface with Opto 22 I/O, radio modems, cellular modems, even satellite communications equipment, as well as any third-party serial device. The SNAP-LCM4 includes built-in diagnostics, a 32-bit Motorola processor, 4 MB RAM, and four expansion slots.
The SNAP-LCSX and SNAP-LCSX-PLUS controllers are the cost-effective answer to applications requiring small, powerful, real-time industrial control. Both contain 32-bit processors capable of a wide range of computing functions. The SNAP-LCSX has two COM ports and the SNAP-LCSX-PLUS has four. Each controller has 1 MB of RAM.
SNAP controllers are programmed using Opto 22 FactoryFloor, an integrated suite of industrial control software applications designed to help you solve control automation problems, build easy-to-use operator interfaces, and expand your manufacturing systems’ connectivity.