Opto 22 Z240D10 DC Control SSR, 240 VAC, 10 A
Representative image only.
Opto 22 Mfr # Z240D10 – Item # EW-68474-34
SPECS & DESCRIPTION
Specifications & Description
  • Load Rating240 VAC
  • Max Current (amps)10
  • DescriptionDC Control SSR, 240 VAC, 10 A
More About this Item
Z Series SSRs are economical 10-amp relays in all-plastic packaging that is made possible through a unique heat transfer system. All Z Series SSRs provide 4000 VRMS of optical isolation and provide a wide operating temperature range of -40°C to +100°C. Z Series SSRs are TTL compatible, with a 3-32 VDC control voltage. All Z Series SSRs feature a built-in snubber and a maximum zero voltage turn-on time of 1/2 cycle. All models are UL recognized and CSA certified.