ORIGIN 6.0 Software, Origin for Macintosh, System 7.5.5, PowerPC
Representative image only.
Mfr # ORIGIN 6.0 – Item # EW-82220-10
Flexible Graphing and Analysis
SPECS & DESCRIPTION
Specifications & Description
  • Operating PlatformMac
  • DescriptionSoftware, Origin for Macintosh, System 7.5.5, PowerPC
More About this Item
Ask about the new Origin 6.0. Graph and analyze data faster and easier than ever before. Choose from a vast array of 2-D and 3-D graph types. Analysis tools include Peak Finding, Peak Fitting, Linear Regression and Nonlinear Curve Fitting with 200 built-in functions. Get powerful math functions including basic statistics, data smoothing, dataset math, integration/differentiation and FFTs. Edit any graphical object with a double-click. Dataset size is limited only by available memory. Open Excel(R) workbooks within Origin. Arrange graphs, worksheets and text in a layout page for publication. Organize data, analysis and results with Project Explorer. Link final results to presentations with OLE 2. Import ASCII, DIF, Sound, LabTech, and more. Export Acrobat, PhotoShop, EPS, TIF, JPEG, and many more. For Macintosh(R).