PacificTransducer 470 Test Stand for Model 411 Type OO Durometer
Representative image only.
PacificTransducer Mfr # 470 – Item # EW-59792-57
Test stands provide the ASTM recommended force for use with durometers
  • 1 year warranty
PRODUCT OPTIONS
$1,719.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DisplayAnalog dial with rotational tare ring
  • DescriptionTest Stand for Model 411 Type OO Durometer
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Compatible for use with the Pacific Transducer Hardness Testers (Durometers), sold separately.