PacificTransducer 473 Deadweight Stand 1KG- 5KG Fortype:A B C D Do And O
Representative image only.
PacificTransducer Mfr # 473 – Item # EW-59792-58
Features an oil-less airpot that gives a smooth, controlled descent for making the most accurate readings with an ASTM Type A, B, or O Durometer
  • Deadweight Stands
  • Load Cell Stand
  • ConstaLoader™ Controlled Descent Stand
  • Maximize Durometer Repeatability
  • Test materials up to 5" high, 3" high for ConstaLoader™
$2,000.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDeadweight Stand 1KG- 5KG Fortype:A B C D Do And O
Key Features
  • Deadweight Stands
  • Load Cell Stand
  • ConstaLoader™ Controlled Descent Stand
  • Maximize Durometer Repeatability
  • Test materials up to 5" high, 3" high for ConstaLoader™
More About this Item
The Deadweight stands use 400G, 1KG, or 5KG weights as recommended by ASTM D2240 specifications. The load cell stand approximates both the 1K & 5Kg weights using calibrated springs. ASTM D2240 states that better repeatability may be obtained by using mass centered on the axis of the indenter. Our stands are designed to give the user maximum repeatability. It also has an adjustable table that can be locked in place for testing materials with nonparallel surfaces.