Parker Hannifin 10RU6-316 Reducing union, 5/8" x 3/8", Stainless Steel, Each
Representative image only.
Parker Hannifin Mfr # 10RU6-316 – Item # EW-30486-14
$89.90USD / Each
BUY MORE & SAVE
  • 1-9$89.90USD / Each
  • 10-49$80.91USD / Each
  • 50-99$76.42USD / Each
  • 100+$71.92USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)5/8
  • Compression Size / Tube Od (Port 2) (in)3/8
  • DescriptionReducing union, 5/8" x 3/8", Stainless Steel, Each
More About this Item
Very good chemical resistance. Rated to 220 psi maximum at 77ºF ( 25ºC). Usable from 32 to 212ºF (0 to 100ºC).