Parker Hannifin 6EE6-316 Compression union elbow, 3/8", Stainless Steel, Each
Representative image only.
Parker Hannifin Mfr # 6EE6-316 – Item # EW-30486-20
$39.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting Material316 Stainless Steel
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)3/8
  • Compression Size / Tube Od (Port 2) (in)3/8
  • DescriptionCompression union elbow, 3/8", Stainless Steel, Each
More About this Item
Very good chemical resistance. Rated to 220 psi maximum at 77ºF ( 25ºC). Usable from 32 to 212ºF (0 to 100ºC).