Parker Hannifin Ball Valve, Polypropylene, Union Connect 1/4'', 1/4'' push-to-connect tube
Representative image only.
Parker Hannifin Mfr # PP4VUC4-MG – Item # EW-01379-61
Create a secure and easy push-to-connect connection
  • Glass reinforced polypropylene (PP) body with a polysulfone ball and EPDM seals
  • NSF 51 listed
  • FDA compliant for contact with food
PRODUCT OPTIONS
$16.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4" Push to Connect
  • Connection TypePush to Connect x Push to Connect
  • Inlet Port1/4" Push to Connect
  • Orifice (in)0.1875
  • Orifice (mm)4.76
  • Wetted MaterialsPolypropylene
  • DescriptionBall Valve, Polypropylene, Union Connect 1/4'', 1/4'' push-to-connect tube
Key Features
  • Glass reinforced polypropylene (PP) body with a polysulfone ball and EPDM seals
  • NSF 51 listed
  • FDA compliant for contact with food
More About this Item
These Parker Hannifin polypropylene (PP) ball valves feature an all plastic design is leak free and corrosion resistant.
Valve, Ball, Polypropylene, Union connector, 1/4'' push to connect tube, 1/4'' push to conect tube, 1/each