Parker Hannifin Compression union elbow, PFA, 1/4"
Representative image only.
Parker Hannifin Mfr # GEC-4 – Item # EW-31320-26
$49.75USD / Each
BUY MORE & SAVE
  • 1-9$49.75USD / Each
  • 10-49$44.78USD / Each
  • 50-99$42.29USD / Each
  • 100+$39.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)1/4
  • Compression Size / Tube Od (Port 2) (in)1/4
  • DescriptionCompression union elbow, PFA, 1/4"
Certificate of Compliance
More About this Item
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.