Parker Hannifin Fitting, PFA, Straight, Compression Bulkhead Union, 1/8" OD
Representative image only.
Parker Hannifin Mfr # GSC-2-P – Item # EW-31320-82
Get a secure seal at higher pressures
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
PRODUCT OPTIONS
Free Certificate of Comliance
$81.50USD / Each
BUY MORE & SAVE
  • 1-9$81.50USD / Each
  • 10-49$73.35USD / Each
  • 50-99$69.28USD / Each
  • 100+$65.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 2) (in)1/8
  • DescriptionCompression bulkhead union, PFA, 1/8
Certificate of Compliance
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Easily install compression fittings to rigid tubing.