Parker Hannifin Fitting, PFA, Straight, Compression to Threaded Adapter, 1/8" OD x 1/8" NPT(M)
Representative image only.
Parker Hannifin Mfr # GAMS-22 – Item # EW-31320-07
Superior chemical resistance with durable PFA
  • Excellent choice to connect rigid tubing systems
  • Leak tight seal allows for less operation downtime
  • Ideal for general-purpose and single-use applications
PRODUCT OPTIONS
Free Certificate of Comliance
$47.00USD / Each
BUY MORE & SAVE
  • 1-9$47.00USD / Each
  • 10-49$42.30USD / Each
  • 50-99$39.95USD / Each
  • 100+$37.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeCompression x NPT(M)
  • Thread size NPT(M) (port 2) (in)1/8
  • Compression Size / Tube Od (Port 1) (in)1/8
  • DescriptionFitting, PFA, Straight, Compression to Threaded Adapter, 1/8" OD x 1/8" NPT(M)
Certificate of Compliance
Key Features
  • Excellent choice to connect rigid tubing systems
  • Leak tight seal allows for less operation downtime
  • Ideal for general-purpose and single-use applications
More About this Item
These Parker Hannifin PFA compression to male thread adapter fittings are ideal for use in corrosive environments and chemical applications. Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.