Parker Hannifin Fitting, Stainless Steel, Straight, Compression Union, 3/4" OD"
Representative image only.
Parker Hannifin Mfr # 12SC12-316 – Item # EW-30486-06
Get a secure seal at higher pressures
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$101.50USD / Each
BUY MORE & SAVE
  • 1-9$101.50USD / Each
  • 10-49$91.35USD / Each
  • 50-99$86.28USD / Each
  • 100+$81.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting Material316 Stainless Steel
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeCompression
  • Compression Size / Tube Od (Port 1) (in)3/4
  • Compression Size / Tube Od (Port 2) (in)3/4
  • DescriptionCompression Union, 1/16" x 3/4", Stainless Steel, Each
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.