Parker Hannifin Flare reducing union, 1/2" x 1/4", PFA, Each
Representative image only.
Parker Hannifin Mfr # FSCR-84 – Item # EW-30490-10
Leak-tight and high-pressure connection for corrosive chemical applications
  • Constructed of durable PFA and PVDF
PRODUCT OPTIONS
$106.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Connection TypeFlare
  • Compression Size / Tube Od (Port 1) (in)1/2
  • Compression Size / Tube Od (Port 2) (in)1/4
  • DescriptionFlare reducing union, 1/2" x 1/4", PFA, Each
Key Features
  • Constructed of durable PFA and PVDF
More About this Item
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.