Parker Hannifin Threaded Reducing Bushing, 1/4" NPT(M) x 1/8" NPT(F), 316 SS
Representative image only.
Parker Hannifin Mfr # 4-2 RB-SS – Item # EW-08539-89
Maintain a permanent connection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$28.75USD / Each
BUY MORE & SAVE
  • 1-9$28.75USD / Each
  • 10-49$25.88USD / Each
  • 50-99$24.44USD / Each
  • 100+$23.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Connection TypeNPT(M) x NPT(F)
  • Thread size NPT(M) (port 1) (in)1/4
  • Thread size NPT(F) (port 2) (in)1/8
  • DescriptionThreaded Reducing Bushing, 1/4" NPT(M) x 1/8" NPT(F), 316 SS
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Threaded fittings offer more permanent installation. Choose the size and material that meets your application needs.