Parker Hannifin XP09T Ultrapure Filter Element, 10 cfm
Representative image only.
Parker Hannifin Mfr # XP09T – Item # EW-29585-02
Filter particles from high-purity gases down to 0.01 micron – 99.99999% efficiency
  • Pleated, borosilicate microfiber element with 96% void volume offers high solids holding capacity
  • Element features stainless steel mesh and endcaps with EPDM seals
  • Collar-style housing offers easy element access without interruption to line
$328.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPVC
  • Micron Rating0.01
  • Flow Rate10 scfm
  • Port Size0.25" NPT(F)
  • DescriptionUltrapure Filter Element, 10 cfm
Key Features
  • Pleated, borosilicate microfiber element with 96% void volume offers high solids holding capacity
  • Element features stainless steel mesh and endcaps with EPDM seals
  • Collar-style housing offers easy element access without interruption to line
More About this Item
These filters are ideal for applications with high-purity gases such as argon, helium, or nitrogen. With an RA.8 ultra-polished surface finish, filter housing is ideal for use in pharmaceutical, aerospace, and semiconductor gas applications. The filter element is a pleated, four-layer borosilicate microfiber; the housing, guards, and endcaps are 316 stainless steel. Element features EPDM seals with a double o-ring click-lock design to ensure filter integrity.