PCO-Tech CAL-LIGHT 400 Light Meter with NIST Calibration Certificate
Representative image only.
PCO-Tech Mfr # CAL-LIGHT 400 – Item # EW-86535-05
SPECS & DESCRIPTION
Specifications & Description
  • Min Measurement (Fc)0.1
  • Max Measurement (Lux)40000
  • Accuracy±5% of reading
  • Display TypeLCD (backlit optional)
  • SensorSilicon photodiode with photopic filter
  • Power9 V Battery (1)
  • DescriptionCal-Light 400 Light Meter with NIST Certificate
NIST Traceable Calibration
More About this Item
This user friendly unit is a calibrated precision light meter capable of measuring ambient illumination. This ergonomically designed unit has an angled sensor with a silicon photodiode, allowing the measurement to be taken and the output to be read simultaneously. With single button operation, a single click switches the unit ''ON'' and holding the button ''HOLDS'' the reading. The light meter automatically shuts off after about one minute.