PCRmax DNA Dekkera bruxellensis, with mastermix
Representative image only.
PCRmax Item # EW-93940-02
Quickly and accurately detect the presence of Dekkera bruxellensis
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedYes
  • DescriptionDekkera bruxellensis, with mastermix
Includes
Lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Get accurate results faster and easier with qPCR. While D. bruxellensis is an important contributor to flavor development in beer, it can produce undesirable aromas in wine that can cause enormous economic losses. Select a complete kit with mastermix or a kit without mastermix and provide your own.