PCRmax PCR Plate 96-Well clear, low profile, half skirt, raised rim, 50/cs
Representative image only.
PCRmax Item # EW-67104-27
Ultrathin walls provide faster heat transfer and cycle times
  • Constructed of high-quality polypropylene for optimal clarity and solvent resistance
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Choose from 96- and 384-well plates
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)0.2
  • MaterialPP
  • DescriptionPCR Plate 96-Well clear, low profile, half skirt, raised rim, 50/cs
Key Features
  • Constructed of high-quality polypropylene for optimal clarity and solvent resistance
  • Certified free of RNase, DNase, endotoxins, and human DNA
  • Choose from 96- and 384-well plates
More About this Item
The transparent 96- and 384-well plates are designed with ultra-thin walls offering accelerated heat transfer and quicker cycle times. The 96-well plates have a well volume of 0.2 mL, are available with full, half, or no skirt, and come with a low or standard profile well. The 384-well plate has a well volume of 40 µl, a sample volume of 25 µl, and features a full skirt. The plates are compatible with most thermal cyclers and are autoclavable.
Not compatible with PCRmax Alpha Cyclers or Techne Thermal Cyclers.