This item is now discontinued. Please browse similar items
PCRmax PCR Strip Tube Caps, Domed, Blue; 125/Cs
Representative image only.
PCRmax Item # EW-67104-19
  • Constructed of pure high-quality polypropylene without additives
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
  • Available in five color options
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • AutoclavableYes
  • Tube ColorBlue
  • Cap StyleDomed
  • Cap MaterialPolypropylene
  • DescriptionPCR Strip Tube Caps, Domed, Blue; 125/Cs
Key Features
  • Constructed of pure high-quality polypropylene without additives
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
  • Available in five color options
More About this Item
Choose between domed and flat-style caps in five color options: clear, blue, red, yellow, or green.