PendoTech Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
Representative image only.
PendoTech Mfr # PDKT-105-03 – Item # EW-19406-35
Optimize performance of your PendoTECH® Pressure Sensor
  • Choose from a wide range of replacement parts and accessories
PRODUCT OPTIONS
$13.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (PSI)76
  • Process Connection1/2" x 1/2" hose barb, male luer
  • DescriptionAdapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
Key Features
  • Choose from a wide range of replacement parts and accessories
More About this Item
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Connects flow through tubing to luer fitting single use pressure sensors.