PendoTech PMAT-CB-MF25 Pressure Mat Interface Cable; 6 ft (1.8 m)
Representative image only.
PendoTech Mfr # PMAT-CB-MF25 – Item # EW-19406-87
Optimize performance of your PendoTECH® Pressure Sensor
  • Choose from a wide range of replacement parts and accessories
PRODUCT OPTIONS
$131.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)72
  • Length (cm)182.88
  • DescriptionInterface cable 6 ft (1.8 m)
Key Features
  • Choose from a wide range of replacement parts and accessories
More About this Item
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
Use this cable to connect to your PendoTECH PressureMAT™ data output to USB serial port.