PendoTech Single-Channel Temperature Sensor Benchtop Transmitter, 4-20 mA Output, ABS Plastic Box; 24VDC
Representative image only.
PendoTech Mfr # PDKT-TT1 – Item # EW-19406-47
Optimize performance of your PendoTECH® Pressure Sensor
  • Choose from a wide range of replacement parts and accessories
  • Produces a 4-20 mA signal that is linear with temperature
PRODUCT OPTIONS
$1,014.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature Sensor System Accessory with 4-20mA Output in ABS Plastic Box; 24VDC Wall Supply ***Must order cable separately***
Two year warranty
Includes
Power cord and plug.
Key Features
  • Choose from a wide range of replacement parts and accessories
  • Produces a 4-20 mA signal that is linear with temperature
More About this Item
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
The single-channel temperature sensor benchtop transmitter easily connects to the PendoTECH temperature sensor. It features a rugged ABC plastic enclosure and LED front panel power indicator. Electrical connections are located on the back panel. The narrow 4-20mA output range from 0 to 70ºC ensures accurate and drift free linearization of the sensor output.
***Must order reusable temperature sensor cables (19406-14 for luer sensor and 19406-15 for hose barb sensors) separately.***