PendoTech TT1 Temperature Sensor Transmitter with 4-20 mA Output
Representative image only.
PendoTech Mfr # TT1 – Item # EW-19406-45
Optimize performance of your PendoTECH® Pressure Sensor
  • Choose from a wide range of replacement parts and accessories
  • Produces a 4-20 milliamp signal that is linear with temperature
  • Connections to the transmitter are quick and convenient
  • Convenient mounting screws
PRODUCT OPTIONS
$368.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature Sensor Transmitter with 4-20mA Output
Two year warranty
Key Features
  • Choose from a wide range of replacement parts and accessories
  • Produces a 4-20 milliamp signal that is linear with temperature
  • Connections to the transmitter are quick and convenient
  • Convenient mounting screws
More About this Item
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
The temperature sensor transmitter easily connects to the PendoTECH temperature sensor. The narrow 4-20mA output range from 0 to 70ºC ensures accurate and drift free linearization of the sensor output.