Pentair 156025-02 304 stainless steel housing for 4 filters
Representative image only.
Pentair Mfr # 156025-02 – Item # EW-29810-05
Stack up to 20 double open end cartridges in one housing!
  • Use standard double-open-end cartridges for high-flow applications
  • Easy to install and maintain with wide-top access port
$1,893.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Material304 Stainless Steel
  • Housing Material21
  • Flow Rate28 GPM
  • Number of Cartridges4
  • Port Size2" NPT(M)
  • Max Temperature (° F)300
  • Max Temperature (° C)149
  • Housing Height (in)53.34
  • Housing Outside Diameter (OD) (in)12
  • Housing Outside Diameter (OD) (cm)30.48
  • Description304 stainless steel housing for 4 filters
Key Features
  • Use standard double-open-end cartridges for high-flow applications
  • Easy to install and maintain with wide-top access port
More About this Item
Stainless steel housings are great for drinking water applications in schools, restaurants, institutions, and industrial plants. Housings hold from 4 to 20 double open end cartridges--filter a large volume of water every minute! Housings feature durable 304 stainless steel construction with epoxy finish to provide better chemical compatibility. All ports are clearly labeled for easy installation.
Maximum pressure is 125 psi; maximum temperature is 300°F (149<>degC). Clean and dirty drains are 1/4", inlet and outlet ports are 2" NPT (M). Order cartridges separately. Buna N seal.