Pentair 257440-06 Quick-Change Filtration System Bracket, Triple-Head, 1/4" OD
Representative image only.
Pentair Mfr # 257440-06 – Item # EW-01507-93
Optimize your Pentair Quick-Change Filtration Systems
  • Choose from replacement cartridges and bracketed heads for your your Pentair Quick-Change Filtration Systems
PRODUCT OPTIONS
$235.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size1/4" OD John Guest® push-to-connect
  • DescriptionQuick-Change Filtration System Bracket, Triple-Head, 1/4" OD
Key Features
  • Choose from replacement cartridges and bracketed heads for your your Pentair Quick-Change Filtration Systems
More About this Item
The quick-change filtration systems offer a complete solution in one simple design—several cartridges will meet your water quality needs. Easily change the cartridge filter with a quarter-turn of the filter. The filter head features a built-in auto shut-off when cartridge is removed, eliminating the need for additional shutoff valves in your filter system. Connections on inlets and outlets are available with 1/4" NPT fittings, or 1/4" and 3/8" John Guest® push-to-connect fittings.
Each quick-change system includes a cartridge, filter head, and mounting bracket for use as a single unit; single cartridge systems are NSF certified. You can also combine cartridges in a series of up to three filters with the optional double and triple bracketed heads (sold separately).