Pentair Double open end hexametaphosphate crystal cartridge; 10"L
Representative image only.
Pentair Mfr # 155251-03 OR 155251-43 – Item # EW-29830-60
Prevents scale and rust buildup—ideal for water processing
$47.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaHexametaphosphate
  • Flow Rate1.5 to 2.5 GPM
  • Filter Length (in)10
  • Max Temperature (° F)100
  • Max Temperature (° C)38
  • DescriptionDouble open end hexametaphosphate crystal cartridge; 10"L
More About this Item
Hexametaphosphate Crystal Cartridge prevents scale and rust buildup — use with water heaters, air conditioning equipment, and many other types of water processing equipment. Food-grade hexametaphosphate dissolves slowly in water to treat scale, corrosion, and iron problems at various flow rates and feed concentrations. All materials are FDA-compliant.
Water being treated should have no more than 257 mg/L hardness, with a pH range of 6.5 to 9.0. Maximum temperature is 100°F (38°C). Cartridge is made from polypropylene (PP) shell with a polyester pad prefilter and a PP post filter.
Live chat
Alternatives
Alternatives