This item is now discontinued. We recommend the following as suggested replacements: EW-78905-17
Polaroid Microscope camera; takes 3 1/4" x 4 1/4" photos
Representative image only.
Mfr # MF-10 – Item # EW-03907-10
Make permanent 3 1/4" x 4 1/4" visual records for comparison and study right in your classroom or laboratory
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPolaroid Microscope camera; takes 3 1/4\" x 4 1/4\" photos
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This camera is compatible with all Cole-Parmer microscopes! Get instant processing—simply load the camera with either Polaroid type 667 black-and-white or type 669 color film (order from table below). Use the focusing tube (included) for fine adjustments, slip the camera on the microscope, and then click the shutter. Seconds later, peel off the picture to reveal a photograph.
Use the universal instrument adapter (included) to fit the camera on virtually any microscope with an eyepiece OD of 34 mm or less (model 48401-10 pictured is ideal). Camera case is constructed of high-impact plastic with a stainless steel film processing chamber. Camera comes complete with sturdy universal instrument adapter, focusing tube, and cable release.