PolyScience HBATH Histology Bath, -76F/-60C, 240 VAC

Representative image only.
PolyScience Mfr # HBATH-60-A12E – Item # EW-10123-05
Freeze samples quickly, preventing ice crystal development and sample distortion
  • Reaches minimum temperature in about one hour
  • Fast-freeze samples with continuous cooling to -76°F (-60°C)
  • Immerse samples easily using included basket and chuck holder
  • Perform maintenance quickly and efficiently via the front-mounted reservoir drain
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Bath TypeCooling
  • Reservoir Capacity (L)1.9
  • Max Temperature (° C)-60
  • Min Temperature (° C)-60
  • Width (in)11
  • Length (in)22 3/4
  • Height (in)11
  • Bath Opening Width (in)4 51/64
  • Port Connections7/16\" (11.5 mm) hose barb
  • Power (VAC)230
  • Power (Hz)50
  • DescriptionHistology Bath, -76F/-60C, 240 VAC
  • Warranty2 Years
Two year warrantyConformite Europeenne

Key Features

  • Reaches minimum temperature in about one hour
  • Fast-freeze samples with continuous cooling to -76°F (-60°C)
  • Immerse samples easily using included basket and chuck holder
  • Perform maintenance quickly and efficiently via the front-mounted reservoir drain
  • 2 year warranty

More About this Item

Histology baths are ideal for freezing samples for enzyme studies. The stainless steel bath reservoir and cover resist corrosion and provide durability. This bath also features easy one button operation, and the DuraTop™ insulating top deck is comfortable to the touch during operation.

Note: Using flammable isopentane as the bath fluid, operation in a fume hood is advised. Please see our selection of Fume Hoods or contact our Application Specialists for assistance.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat