Pomona Electronics 6497 Modular Passive Oscilloscope Probe, 10:1 Ratio
Representative image only.
Pomona Electronics Mfr # 6497 – Item # EW-20009-51
Available for sale in the United States and Canada only
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Bandwidth (MHz)350
  • DescriptionModular Passive Oscilloscope Probe, 10:1 Ratio
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Featuring a new, compact, ergonomicallydesigned probe handle, these modular general purpose passive voltage probes are ideal for use with a wide variety of oscilloscopes. They also feature a spring-loaded, pogo-pin replaceable tip.The fine tip probes are perfectly suited for probing high-density chip leads with less than 0.060 in(1.52mm) spacing.
Modular in design, the probe handle is capable of receiving a variety of spring-loaded and fixed tips, providing greater testing versatility. An affordable,versatile probe for many uses.
Probe sets feature probe handle, insulated adjusting tool, sprung hook, BNC adapter, 9.06 in (230mm) ground lead, 0.98 in (25mm) ground adapter, IC tip adapter, tip cover, rigid tip and spring-loaded tip (1 each)