Pomona Electronics 72934 Multi-RF Direct Adapter Kit
Representative image only.
Pomona Electronics Mfr # 72934 – Item # EW-23031-92
  • All 26 adapters in kit are direct.
  • No intermediate couplers
  • Reduced signal loss in adapters due to absence of couplers
  • Kit includes the most frequently used adapters including BNC/BNC, N/N, SMA/SMA, BNC/N,
  • SMA/BNC, SMA/N, SMA/TNC
  • Rugged, lockable, pistol-grip case
  • All parts precision machined brass with tarnish resistant gold or nickel plating
  • Male center pins are gold plated brass
  • Female contacts are gold plated heat treated beryllium copper
  • Insulation material is high density PTFE
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMulti-RF Direct Adapter Kit
Key Features
  • All 26 adapters in kit are direct.
  • No intermediate couplers
  • Reduced signal loss in adapters due to absence of couplers
  • Kit includes the most frequently used adapters including BNC/BNC, N/N, SMA/SMA, BNC/N,
  • SMA/BNC, SMA/N, SMA/TNC
  • Rugged, lockable, pistol-grip case
  • All parts precision machined brass with tarnish resistant gold or nickel plating
  • Male center pins are gold plated brass
  • Female contacts are gold plated heat treated beryllium copper
  • Insulation material is high density PTFE