Precision Digital Explosion-Proof Rate/Totalizer, 9-30 VDC, with backlight.

Representative image only.
Precision Digital Mfr # PD6830-AP0-0 – Item # EW-30004-63
Program your process meter on-site
  • 5-digit rate and 7-character total/tag display
  • Backlight standard
  • Rotatable display
  • FM, CSA, ATEX, IECEx, CE approved

PRODUCT OPTIONS

$859.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Input TypePulse; open collector (NPN, PNP, TTL); switch contact
  • Display Type0.7" LED; 0.4" LED
  • Display Digits5; 7
  • Max Power VDC (VDC)30
  • Min Power VDC (VDC)9
  • Max Operating Temperature (° F)167
  • Min Operating Temperature (° F)-40
  • EnclosureNEMA 4X, 7, 9; IP68; explosion-proof die-cast aluminum
  • Overall Width (in)5
  • Overall Height (in)5 1/4
  • Overall Depth (in)4
  • Overall Width (mm)142.9
  • Overall Height (mm)133.4
  • Overall Depth (mm)123.8
  • DescriptionExplosion-Proof Rate/Totalizer, 9-30 VDC, with backlight.

Key Features

  • 5-digit rate and 7-character total/tag display
  • Backlight standard
  • Rotatable display
  • FM, CSA, ATEX, IECEx, CE approved

More About this Item

The ProtEX-RTs are rugged, explosion-proof, full-featured rate/totalizers designed for demanding applications in hazardous areas or in the harshest environmental conditions. The PD6820 accepts analog inputs and the PD6830 accepts pulse inputs. They are programmed using the four SafeTouch™ through-glass buttons without removing the cover, or the four mechanical buttons. The sleek die-cast aluminum enclosure is provided with two ¾-inch threaded conduit holes and integrated pipe or wall mounting flanges.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat