Prepared Media Bottles, saline, 0.45%, 1 L bottle, 10/pk
Representative image only.
Item # EW-14204-86
Save time and money with ready-made bottles
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For the preparation of isotonic suspensions and dilutions of microorganisms
PRODUCT OPTIONS
$131.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Saline
  • Capacity (mL)1000
  • Capacity (L)1
  • DescriptionPrepared Media Bottles, saline, 0.45%, 1 L bottle, 10/pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For the preparation of isotonic suspensions and dilutions of microorganisms
More About this Item
Bottles are tested and standardized to ensure consistent and reliable performance. Each bottle includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Bottle and pack sizes vary.