Prepared media tube, Acetate differential—prepared slant tube, 20 pack
Representative image only.
Item # EW-14203-52
Save time and money with ready-made tubes
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For the differentiation of E. coli and Shigella spp
  • Tube size: 16 x 100 mm
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Acetate Differential
  • DescriptionPrepared media tube, Acetate differential—prepared slant tube, 20 pack
  • Warranty1 year
1 Year Warranty
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For the differentiation of E. coli and Shigella spp
  • Tube size: 16 x 100 mm
  • 1 year warranty
More About this Item
Tubes are tested and standardized to ensure consistent and reliable performance. Each tube includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Tube and pack sizes vary.