Prepared media tube, Blood agar, 5%—prepared slant tube, 20 pack
Representative image only.
Item # EW-14203-58
Save time and money with ready-made tubes
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • Tryptic soy agar with 5% sheep blood for the cultivation of microorganisms
  • Tube size: 16 x 100 mm
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Blood
  • DescriptionPrepared media tube, Blood agar, 5%—prepared slant tube, 20 pack
  • Warranty3 month
3 Month Warranty
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • Tryptic soy agar with 5% sheep blood for the cultivation of microorganisms
  • Tube size: 16 x 100 mm
  • 3 month warranty
More About this Item
Tubes are tested and standardized to ensure consistent and reliable performance. Each tube includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Tube and pack sizes vary.