Prepared Slant Media Tubes, BHI Agar w/5% Sheep's Blood; 20/Pk
Representative image only.
Item # EW-14203-56
Save time and money with ready-made tubes
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • Brain heart infusion agar with 5% sheep blood for cultivation of fungi
  • Tube size: 20 x 125 mm
PRODUCT OPTIONS
$194.00USD / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Brain-Heart Infusion w/ 5% Sheep's Blood
  • DescriptionPrepared Slant Media Tubes, BHI Agar w/5% Sheep's Blood; 20/Pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • Brain heart infusion agar with 5% sheep blood for cultivation of fungi
  • Tube size: 20 x 125 mm
More About this Item
Tubes are tested and standardized to ensure consistent and reliable performance. Each tube includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Tube and pack sizes vary.