Ralston Instruments QTHA-2FS0-QD 1/4" female NPT Quick-connect x male Quick-test, S.S.
Representative image only.
Ralston Instruments Mfr # QTHA-2FS0-QD – Item # EW-68976-88
Adapters have a unique sealing system that makes them excellent for both high and low pressure calibration.
  • ABS case with foam cut-outs makes it easy to see if any adapters are missing
  • Adapters are available in kits or individually.
  • 1 year warranty
PRODUCT OPTIONS
$88.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure5000 psi (345 bar)
  • Min Pressure0 psi (0 bar)
  • Description1/4\" female NPT Quick-connect x male Quick-test, S.S.
  • Warranty1 Year
1 Year Warranty
Key Features
  • ABS case with foam cut-outs makes it easy to see if any adapters are missing
  • Adapters are available in kits or individually.
  • 1 year warranty
More About this Item
Kits take the guesswork out of selecting adapters by providing a selection of the most popular combinations of hoses and adapters necessary to connect to any pressure transmitter, gauge, hand pump, manifold or nitrogen source. Use the individual hoses and fittings to connect to any device you will be calibrating.