Pressure Relief Valve, 316 SS, 1/2" NPT(F)
Representative image only.
Item # EW-03245-40
Easily control your outlet pressure
  • Constructed of durable 316 stainless steel (SS)
$819.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection TypeNPT(F)
  • Orifice (in)0.3125
  • Orifice (mm)7.9375
  • Max Pressure (PSI)150
  • Wetted MaterialsStainless Steel
  • DescriptionPressure Relief Valve, 316 SS, 1/2" NPT(F)
Key Features
  • Constructed of durable 316 stainless steel (SS)
More About this Item
Use for liquid applications with flow rates up to 15 GPM. For pressures from 40 to 150 psig. Valves have 1/2" NPT(F); orifice size is 5/16".