PrimeLock® Clamp Insert, EPDM-lined PFA, standard, 1/2" OD tubing; 2/set
Representative image only.
Item # EW-30493-39
Optimize performance of your PrimeLock® system
  • Choose from a wide range of replacement parts and accessories
PRODUCT OPTIONS
$276.00USD / Set of 2
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialEPDM-lined PFA
  • Connection TypeFlare Clamp Insert
  • Compression Size / Tube Od (Port 1) (in)1/2
  • DescriptionClamp Insert, EPDM-lined PFA, standard, 1/2" OD tubing; 2/set
Key Features
  • Choose from a wide range of replacement parts and accessories
More About this Item
Installation tools require mandrel(s) and clamp insert(s) for operation. EPDM-lined clamp inserts eliminate surface marks on tubing following insert installation. Minimum tube union installations require the appropriately sized minimum tube union mandrel in addition to the standard installation mandrel. Directional sleeve mandrels prevent backwards installation of the insert, sizes 3/8" to 1-1/4".