PrimeLock® High-Purity Fitting, nut, standard, 1/4" OD tubing; 1/ea
Representative image only.
Item # EW-30492-04
  • Replacement nuts for use with any PrimeLock PFA fitting
PRODUCT OPTIONS
$11.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Connection TypeFlare
  • Compression Size / Tube Od (Port 1) (in)1/4
  • ColorNatural
  • Min Temperature (° F)0
  • Max Temperature (° F)392
  • Min Temperature (° C)-18
  • Max Temperature (° C)200
  • Max Operating Pressure (PSI)120 at 70°F (21°C)
  • DescriptionHigh-Purity Fitting, nut, standard, 1/4" OD tubing; 1/ea
Key Features
  • Replacement nuts for use with any PrimeLock PFA fitting
More About this Item
Replacement parts and accessories for PrimeLock® High-Purity Fittings.