PrimeLock® Installation Insert, PFA, 1" OD tubing; 10/pk
Representative image only.
Item # EW-30491-95
Easily install your High-Purity PFA Fittings
  • Optimize performance of your PFA Flare Fittings
  • Replacement nonwetted inserts eliminate entrapment, leak points and flow interruptions
PRODUCT OPTIONS
$144.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Connection TypeFlare Installation Insert
  • Compression Size / Tube Od (Port 1) (in)1
  • ColorNatural
  • Min Temperature (° F)0
  • Max Temperature (° F)392
  • Min Temperature (° C)-18
  • Max Temperature (° C)200
  • Max Operating Pressure (PSI)120 at 70°F (21°C)
  • DescriptionInstallation Insert, PFA, 1" OD tubing; 10/pk
Key Features
  • Optimize performance of your PFA Flare Fittings
  • Replacement nonwetted inserts eliminate entrapment, leak points and flow interruptions
More About this Item
Replacement parts and accessories for PrimeLock® High-Purity Fittings.