PRO Scientific Deflector Head for Homogenizer Probe, 20 mm; Plastic

Representative image only.
PRO Scientific Mfr # 02-20000D – Item # EW-04751-84
Expand and upgrade your homogenizing capabilities

PRODUCT OPTIONS

$334.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialAcetal Resin
  • Compatible WithPRO Scientific Generator Probes, 20 mm diameter
  • Diameter (mm)20
  • DescriptionDeflector Head for Homogenizer Probe, 20 mm; Plastic
Two year warranty

More About this Item

Parts and accessories compatible with PRO Scientific homogenizers.

Deflector Heads enhance homogenization by increasing sample turnover. Ideal for large or viscous samples, the design creates vertical movement of the sample from the generator tip, repeatedly cycling the material up and down. Choose between stainless steel and acetal resin construction.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat