Pulsafeeder L380FT01-PVD Solenoid Metering Pump Five-Function Valve, PVDF, 150 psi, 3/8" OD
Representative image only.
Pulsafeeder Mfr # L380FT01-PVD – Item # EW-74113-86
Optimize performance of your Pulsafeeder Solenoid Metering Pump
  • Choose from a wide range of replacement parts and accessories
PRODUCT OPTIONS
$95.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPVDF five-function valve, compression fitting, 3/8" tubing connectors, 150 psi max
Key Features
  • Choose from a wide range of replacement parts and accessories
More About this Item
Complete and customize a pump system for any application with a variety of valves and mounting options. Service kits are also available to make routine maintenance easy.