Pulsafeeder XP0095ANX Adjustable-Speed Peristaltic Pump; 9GPD/100PSI/230V/50Hz
Representative image only.
Pulsafeeder Mfr # XP0095ANX – Item # EW-74130-73
Easy maintenance and no contamination—tubing is the only wetted part
  • Easy tube change-out minimizes downtime between tubing set changes
  • Inherent degassing function and rugged gear train offer many years of reliable service
PRODUCT OPTIONS
$635.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (mL/min)23
  • Max Pressure (PSI)100
  • Power (VAC)230
  • Power (Hz)50
  • Power Cord6-ft power cord
  • ETL Regulatory ApprovalYes
  • DescriptionAdjustable-Speed Peristaltic Pump; 9GPD/100PSI/230V/50Hz
Includes
pump head, two tubing assemblies, and a 6-ft (1.8-m) power cord.
Key Features
  • Easy tube change-out minimizes downtime between tubing set changes
  • Inherent degassing function and rugged gear train offer many years of reliable service
More About this Item
These simple-to-use, self-priming peristaltic pumps deliver worry-free dosing.