Puma AT-5133 Heavy Duty air impact wrench, 7,000 rpm, 200 ft-lb torque, 1/4 in connection, 90 psi
Representative image only.
Puma Mfr # AT-5133 – Item # EW-07122-03
  • Deliver high torque output with minimal exertion
  • Standard, butterfly, and pistol-grip styles available
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Free Air Capacity (CFM)12
  • Max Pressure (PSI)90
  • DescriptionHeavy Duty air impact wrench, 7,000 rpm, 200 ft-lb torque, 1/4 in connection, 90 psi
  • Warranty1 year
1 Year Warranty
Key Features
  • Deliver high torque output with minimal exertion
  • Standard, butterfly, and pistol-grip styles available
  • 1 year warranty
More About this Item
Make the most of your air compressor with these air-powered tools. All require 90 psi pressure, but vary in air consumption and fittings. A variety of drive sizes, bolt capacities and maximum torque provide you with plenty of options.